Assay Details
Target Gene Details
Entrez Gene ID: | 63036 |
Gene Name: | chymotrypsin like elastase family member 2A |
Gene Aliases: |
ELA2A, PE-1 |
Location: |
Chr.1:15456728-15472091 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CELA2A | NM_033440.2 | NP_254275.1 | ||
| AK312198.1 | ||||
| BC007031.1 | AAH07031.1 | |||
| BC069331.1 | AAH69331.1 | |||
| BC069432.1 | AAH69432.1 | |||
| BP324428.1 | ||||
| CR450291.1 | CAG29287.1 | |||
| D00236.1 | BAA00165.1 | |||
| M16631.1 | AAA52374.1 | |||
| M16652.1 | AAA52380.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2744308 | Chr.1:15455456 - 15475909 on Build GRCh38 | Deletion |
|
| esv3548899 | Chr.1:15454506 - 15475973 on Build GRCh38 | Deletion |
|
| nsv833536 | Chr.1:15323505 - 15528830 on Build GRCh38 | Loss |
|
| nsv1114929 | Chr.1:15455856 - 15475193 on Build GRCh38 | Deletion |
|
| esv2761582 | Chr.1:15457276 - 15475000 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map