Assay Details
Target Gene Details
Entrez Gene ID: | 400746 |
Gene Name: | non-compact myelin associated protein |
Gene Aliases: |
C1orf130, MP11 |
Location: |
Chr.1:24556076-24609328 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCMAP | NM_001010980.4 | NP_001010980.1 | ||
| XM_011541463.2 | XP_011539765.1 | |||
| AK124519.1 | ||||
| CB961010.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545823 | Chr.1:24568506 - 24621457 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79456857] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map