Assay Details
Target Gene Details
Entrez Gene ID: | 7799 |
Gene Name: | PR domain 2 |
Gene Aliases: |
HUMHOXY1, KMT8, MTB-ZF, RIZ, RIZ1, RIZ2 |
Location: |
Chr.1:13698875-13825079 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRDM2 | XM_017002264.1 | 1 | 480 | XP_016857753.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545567 | Chr.1:13558784 - 14026576 on Build GRCh38 | Gain |
|
| nsv3187 | Chr.1:13665844 - 13711151 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map