Assay Details
Target Gene Details
Entrez Gene ID: | 5928 |
Gene Name: | RB binding protein 4, chromatin remodeling factor |
Gene Aliases: |
NURF55, RBAP48, lin-53 |
Location: |
Chr.1:32651148-32686211 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBBP4 | NM_001135255.1 | NP_001128727.1 | ||
| NM_001135256.1 | NP_001128728.1 | |||
| NM_005610.2 | NP_005601.1 | |||
| AK222779.1 | BAD96499.1 | |||
| AK299251.1 | ||||
| AK299410.1 | ||||
| AK312571.1 | ||||
| BC003092.1 | AAH03092.1 | |||
| BC015123.1 | AAH15123.1 | |||
| BC053904.1 | AAH53904.1 | |||
| BC075836.1 | AAH75836.1 | |||
| BT007309.1 | AAP35973.1 | |||
| X71810.1 | CAA50685.1 | |||
| X74262.1 | CAA52321.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545961 | Chr.1:32566811 - 32685350 on Build GRCh38 | Loss |
|
| nsv519188 | Chr.1:32619643 - 32780839 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map