Assay Details
Target Gene Details
Entrez Gene ID: | 56950 |
Gene Name: | SET and MYND domain containing 2 |
Gene Aliases: |
HSKM-B, KMT3C, ZMYND14 |
Location: |
Chr.1:214281101-214337134 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMYD2 | NM_020197.2 | NP_064582.2 | ||
| AF226053.1 | AAF86953.1 | |||
| AK313868.1 | ||||
| BC028104.1 | ||||
| BC098133.1 | AAH98133.1 | |||
| BC098276.1 | AAH98276.1 | |||
| BC098335.1 | AAH98335.1 | |||
| BG723585.1 |
More Information
Set Membership: |
|

Genomic Map