Assay Details
Target Gene Details
Entrez Gene ID: | 9406 |
Gene Name: | zinc finger RANBP2-type containing 2 |
Gene Aliases: |
ZIS, ZIS1, ZIS2, ZNF265 |
Location: |
Chr.1:71063291-71081289 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZRANB2 | NM_005455.4 | NP_005446.2 | ||
| NM_203350.2 | NP_976225.1 | |||
| AB209568.1 | BAD92805.1 | |||
| AF065391.1 | AAD09746.1 | |||
| AF065392.1 | AAD09747.1 | |||
| AK222858.1 | BAD96578.1 | |||
| AL136945.1 | CAB66879.1 | |||
| AU100296.1 | ||||
| BC039814.1 | AAH39814.1 | |||
| DA523601.1 |
More Information
Set Membership: |
|

Genomic Map