Assay Details
Target Gene Details
Entrez Gene ID: | 1600 |
Gene Name: | DAB1, reelin adaptor protein |
Gene Aliases: |
- |
Location: |
Chr.1:56997906-58250539 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DAB1 | NM_021080.3 | NP_066566.3 | ||
| AB210012.1 | ||||
| AK095513.1 | ||||
| CR933698.1 |
Target Gene Details
Entrez Gene ID: | 101926890 |
Gene Name: | DAB1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:57860543-57863114 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DAB1-AS1 | NR_104362.1 | 1 | 306 | |
| NR_104363.1 | 1 | 306 | ||
| NR_104364.1 | 1 | 306 | ||
| NR_104365.1 | ||||
| BU570679.1 | 1 | 258 | ||
| DB458876.1 | 1 | 237 | ||
| DB460815.1 | 1 | 256 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829937 | Chr.1:57792616 - 57940834 on Build GRCh38 | Gain |
|
| esv3893568 | Chr.1:57618490 - 58256638 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map