Assay Details
Target Gene Details
Entrez Gene ID: | 23456 |
Gene Name: | ATP binding cassette subfamily B member 10 |
Gene Aliases: |
EST20237, M-ABC2, MTABC2 |
Location: |
Chr.1:229516582-229558987 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCB10 | NM_012089.2 | NP_036221.2 | ||
| XM_011544135.2 | XP_011542437.1 | |||
| XM_011544136.2 | XP_011542438.1 | |||
| AB013380.1 | BAB20265.1 | |||
| AF216833.1 | AAF78198.1 | |||
| AK172794.1 | BAD18770.1 | |||
| BC064930.1 | AAH64930.1 |
More Information
Set Membership: |
|

Genomic Map