Assay Details
Target Gene Details
Entrez Gene ID: | 252839 |
Gene Name: | transmembrane protein 9 |
Gene Aliases: |
DERM4, TMEM9A |
Location: |
Chr.1:201134771-201171625 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428301 | Chr.1:201059216 - 201191346 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map