Assay Details
Target Gene Details
Entrez Gene ID: | 103021296 |
Gene Name: | uncharacterized LOC103021296 |
Gene Aliases: |
- |
Location: |
Chr.1:205934913-205978095 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC103021296 | NR_110983.1 | |||
| NR_110984.1 | ||||
| AK023966.1 | ||||
| AU142174.1 | ||||
| DA449064.1 |
Target Gene Details
Entrez Gene ID: | 115019 |
Gene Name: | solute carrier family 26 member 9 |
Gene Aliases: |
- |
Location: |
Chr.1:205913048-205943472 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC26A9 | NM_052934.3 | NP_443166.1 | ||
| NM_134325.2 | NP_599152.2 | |||
| XM_011509121.2 | XP_011507423.1 | |||
| XM_011509124.2 | XP_011507426.1 | |||
| AF331525.1 | AAK95667.1 | |||
| BC136538.1 | ||||
| BC151208.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832392 | Chr.1:205882365 - 205944905 on Build GRCh38 | Gain |
|
| nsv470777 | Chr.1:205922508 - 205945757 on Build GRCh38 | Loss |
|
| esv29758 | Chr.1:205925403 - 205953545 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map