Assay Details
Target Gene Details
Entrez Gene ID: | 64754 |
Gene Name: | SET and MYND domain containing 3 |
Gene Aliases: |
KMT3E, ZMYND1, ZNFN3A1, bA74P14.1 |
Location: |
Chr.1:245749340-246507342 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMYD3 | NM_001167740.1 | NP_001161212.1 | ||
| NM_022743.2 | NP_073580.1 | |||
| XM_011544253.1 | XP_011542555.1 | |||
| XM_011544254.2 | XP_011542556.1 | |||
| XM_011544257.1 | XP_011542559.1 | |||
| XM_011544258.2 | XP_011542560.1 | |||
| XM_011544259.2 | XP_011542561.1 | |||
| XM_011544260.1 | XP_011542562.1 | |||
| XM_011544261.2 | XP_011542563.1 | |||
| XM_017002094.1 | XP_016857583.1 | |||
| XM_017002095.1 | XP_016857584.1 | |||
| AB057595.1 | BAB86333.1 | |||
| AK023594.1 | ||||
| AK024733.1 | BAB14981.1 | |||
| AK289605.1 | ||||
| AW173130.1 | ||||
| AY186742.1 | AAO31695.1 | |||
| BC017079.1 | AAH17079.2 | |||
| BC031010.1 | AAH31010.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3589447 | Chr.1:245767201 - 245780520 on Build GRCh38 | Loss |
|
| esv3891326 | Chr.1:245759378 - 245988873 on Build GRCh38 | Gain |
|
| esv3575839 | Chr.1:245526258 - 245999859 on Build GRCh38 | Gain |
|
| nsv998007 | Chr.1:245721457 - 245826060 on Build GRCh38 | Loss |
|
| nsv549520 | Chr.1:245750982 - 245836536 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map