Assay Details
Target Gene Details
Entrez Gene ID: | 57568 |
Gene Name: | signal induced proliferation associated 1 like 2 |
Gene Aliases: |
SPAL2 |
Location: |
Chr.1:232397965-232630491 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIPA1L2 | NM_020808.4 | NP_065859.3 | ||
| XM_005273211.2 | XP_005273268.1 | |||
| XM_005273212.4 | XP_005273269.1 | |||
| XM_005273213.4 | XP_005273270.1 | |||
| XM_011544243.2 | XP_011542545.1 | |||
| XM_017001896.1 | XP_016857385.1 | |||
| XM_017001897.1 | XP_016857386.1 | |||
| XM_017001898.1 | XP_016857387.1 | |||
| AB037810.1 | BAA92627.1 | |||
| AK022852.1 | BAB14273.1 | |||
| AK026779.1 | ||||
| AK074212.1 | ||||
| AK093191.1 | BAC04090.1 | |||
| AY168879.1 | AAO12530.1 | |||
| BC013119.1 | ||||
| BC082251.1 | AAH82251.1 | |||
| BC150334.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006763 | Chr.1:232341238 - 232458244 on Build GRCh38 | Gain |
|
| nsv549355 | Chr.1:232388426 - 232435636 on Build GRCh38 | Gain |
|
| esv2724972 | Chr.1:232418345 - 232420440 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map