Assay Details
Target Gene Details
Entrez Gene ID: | 8659 |
Gene Name: | aldehyde dehydrogenase 4 family member A1 |
Gene Aliases: |
ALDH4, P5CD, P5CDh |
Location: |
Chr.1:18871430-18902799 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH4A1 | NM_001161504.1 | NP_001154976.1 | ||
| NM_001319218.1 | NP_001306147.1 | |||
| NM_003748.3 | NP_003739.2 | |||
| NM_170726.2 | NP_733844.1 | |||
| AK222486.1 | BAD96206.1 | |||
| AK289972.1 | ||||
| AK294552.1 | ||||
| BC007581.1 | AAH07581.1 | |||
| BC023600.2 | AAH23600.1 | |||
| BM856369.1 | ||||
| DC318624.1 | ||||
| DC342814.1 | ||||
| FJ462711.1 | ||||
| U24266.1 | AAC50500.1 | |||
| U24267.1 | AAC50501.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76401761] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map