Assay Details
Target Gene Details
Entrez Gene ID: | 56605 |
Gene Name: | endoplasmic reticulum oxidoreductase 1 beta |
Gene Aliases: |
ERO1LB, Ero1beta |
Location: |
Chr.1:236215122-236282039 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERO1B | NM_019891.3 | NP_063944.3 | ||
| XM_017001834.1 | XP_016857323.1 | |||
| AF252538.1 | AAF97547.1 | |||
| AK293941.1 | ||||
| AK315351.1 | ||||
| BC032823.2 | AAH32823.2 | |||
| BC044573.1 | AAH44573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv590n100 | Chr.1:236139422 - 236267713 on Build GRCh38 | Gain |
|
| nsv1005877 | Chr.1:235912335 - 237038508 on Build GRCh38 | Gain |
|
| nsv522991 | Chr.1:236215849 - 236250146 on Build GRCh38 | Loss |
|
| nsv832969 | Chr.1:236111012 - 236311443 on Build GRCh38 | Gain |
|
| nsv549394 | Chr.1:235876577 - 236629706 on Build GRCh38 | Gain |
|
| nsv1012436 | Chr.1:235860657 - 236648100 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map