Assay Details
Target Gene Details
Entrez Gene ID: | 1604 |
Gene Name: | CD55 molecule (Cromer blood group) |
Gene Aliases: |
CR, CROM, DAF, TC |
Location: |
Chr.1:207321472-207360966 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD55 | NM_000574.4 | 10 | 2552 | NP_000565.1 |
| NM_001114752.2 | 11 | 2670 | NP_001108224.1 | |
| NM_001300903.1 | 11 | 2667 | NP_001287832.1 | |
| NM_001300904.1 | 11 | 2645 | NP_001287833.1 | |
| NR_125349.1 | 11 | 2637 | ||
| AW316643.1 | 1 | 246 | ||
| CA432400.1 | 1 | 261 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4221 | Chr.1:207353427 - 207384212 on Build GRCh38 | Deletion |
|
| nsv508688 | Chr.1:207356868 - 207381104 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map