Assay Details
Target Gene Details
Entrez Gene ID: | 100422212 |
Gene Name: | eukaryotic translation initiation factor 3 subunit J pseudogene |
Gene Aliases: |
- |
Location: |
Chr.1:163421070-163423191 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100422212 | NR_104294.1 | 1 | 485 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831770 | Chr.1:163370623 - 163538890 on Build GRCh38 | Loss |
![]() |
