Assay Details
Target Gene Details
Entrez Gene ID: | 116228 |
Gene Name: | COX20, cytochrome c oxidase assembly factor |
Gene Aliases: |
FAM36A |
Location: |
Chr.1:244835306-244845063 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX20 | NM_001312871.1 | NP_001299800.1 | ||
| NM_001312872.1 | NP_001299801.1 | |||
| NM_001312873.1 | NP_001299802.1 | |||
| NM_001312874.1 | NP_001299803.1 | |||
| NM_198076.5 | NP_932342.1 | |||
| NR_132419.1 | ||||
| NR_132420.1 | ||||
| NR_132421.1 | ||||
| AF086182.1 | ||||
| AK000866.1 | ||||
| AK125259.1 | ||||
| AK130498.1 | ||||
| AK300760.1 | ||||
| BC018519.1 | AAH18519.1 | |||
| BC062419.1 | AAH62419.1 | |||
| BC095486.1 | AAH95486.1 | |||
| BG708903.1 | ||||
| BQ189361.1 | ||||
| DA157561.1 | ||||
| DA164934.1 | ||||
| DA752002.1 | ||||
| DB189037.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549493 | Chr.1:244721983 - 244856983 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map