Assay Details
Target Gene Details
Entrez Gene ID: | 7398 |
Gene Name: | ubiquitin specific peptidase 1 |
Gene Aliases: |
UBP |
Location: |
Chr.1:62436304-62451804 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP1 | NM_001017415.1 | NP_001017415.1 | ||
| NM_001017416.1 | NP_001017416.1 | |||
| NM_003368.4 | NP_003359.3 | |||
| AB014458.1 | BAA34703.1 | |||
| AB208893.1 | BAD92130.1 | |||
| AF117386.1 | AAD11441.1 | |||
| AK312423.1 | ||||
| AL117575.1 | CAB55999.1 | |||
| BC050525.1 | AAH50525.1 |
More Information
Set Membership: |
|

Genomic Map