Assay Details
Target Gene Details
Entrez Gene ID: | 5322 |
Gene Name: | phospholipase A2 group V |
Gene Aliases: |
FRFB, GV-PLA2, PLA2-10, hVPLA(2) |
Location: |
Chr.1:20028350-20091901 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G5 | XM_005245891.4 | XP_005245948.1 | ||
| XM_005245892.4 | XP_005245949.1 | |||
| XM_005245893.4 | XP_005245950.1 | |||
| XM_011541586.2 | XP_011539888.1 | |||
| XM_011541587.2 | XP_011539889.1 | |||
| XM_011541588.2 | XP_011539890.1 | |||
| XM_011541589.2 | XP_011539891.1 | |||
| XM_011541590.2 | XP_011539892.1 | |||
| XM_011541591.2 | XP_011539893.1 | |||
| XM_011541592.2 | XP_011539894.1 | |||
| AK097693.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545731 | Chr.1:19989106 - 20776306 on Build GRCh38 | Gain |
|
| nsv1003802 | Chr.1:20057935 - 20154347 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map