Assay Details
Target Gene Details
Entrez Gene ID: | 83416 |
Gene Name: | Fc receptor like 5 |
Gene Aliases: |
BXMAS1, CD307, CD307e, FCRH5, IRTA2, PRO820 |
Location: |
Chr.1:157513377-157552520 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCRL5 | NM_001195388.1 | NP_001182317.1 | ||
| NM_031281.2 | NP_112571.2 | |||
| XM_011510030.1 | XP_011508332.1 | |||
| XM_011510031.1 | XP_011508333.1 | |||
| XM_011510032.1 | XP_011508334.1 | |||
| XM_011510033.1 | XP_011508335.1 | |||
| AF343662.1 | AAK31325.1 | |||
| AF343663.1 | AAK31326.1 | |||
| AF343664.1 | AAK31327.1 | |||
| AF343665.1 | AAK31328.1 | |||
| AF369794.2 | AAK50059.2 | |||
| AF397453.1 | AAK93971.1 | |||
| AK300010.1 | ||||
| AK301356.1 | ||||
| AL834187.1 | CAH56486.1 | |||
| AY358085.1 | AAQ88452.1 | |||
| BC101066.2 | AAI01067.1 | |||
| BC101067.1 | AAI01068.1 | |||
| BC101069.2 | AAI01070.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2718507 | Chr.1:157533583 - 157635387 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map