Assay Details
Target Gene Details
Entrez Gene ID: | 10451 |
Gene Name: | vav guanine nucleotide exchange factor 3 |
Gene Aliases: |
- |
Location: |
Chr.1:107571160-107964923 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VAV3 | NM_001079874.1 | NP_001073343.1 | ||
| NM_006113.4 | NP_006104.4 | |||
| XM_005270360.2 | XP_005270417.1 | |||
| XM_017000053.1 | XP_016855542.1 | |||
| XM_017000054.1 | 26 | 5501 | XP_016855543.1 | |
| XM_017000055.1 | XP_016855544.1 | |||
| XM_017000056.1 | XP_016855545.1 | |||
| AF067817.1 | AAC79695.1 | |||
| AF118886.1 | AAD20348.1 | |||
| AF118887.1 | AAD20349.1 | |||
| AK296537.1 | ||||
| AK300474.1 | ||||
| AK304088.1 | ||||
| AK316295.1 | ||||
| BC143969.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830993 | Chr.1:107441433 - 107613979 on Build GRCh38 | Gain |
|
| dgv248n100 | Chr.1:107385172 - 108286989 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74714642] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map