Assay Details
Target Gene Details
| Entrez Gene ID: | 55277 | 
| Gene Name: | FGGY carbohydrate kinase domain containing | 
| Gene Aliases: | - | 
| Location: | Chr.1:59296405-59762730 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 3 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv829982 | Chr.1:59247176 - 59367120 on Build GRCh38 | Loss |  FGGY | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top