Assay Details
Target Gene Details
Entrez Gene ID: | 23266 |
Gene Name: | adhesion G protein-coupled receptor L2 |
Gene Aliases: |
CIRL2, CL2, LEC1, LPHH1, LPHN2 |
Location: |
Chr.1:81306155-81992738 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRL2 | XM_017000782.1 | XP_016856271.1 | ||
| AK123422.1 |
Target Gene Details
Entrez Gene ID: | 101927434 |
Gene Name: | uncharacterized LOC101927434 |
Gene Aliases: |
- |
Location: |
Chr.1:81513880-81557702 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927434 | NR_125943.1 | |||
| BX092247.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2667433 | Chr.1:81533928 - 81542519 on Build GRCh38 | Deletion |
|
| esv3586616 | Chr.1:81533884 - 81542659 on Build GRCh38 | Loss |
|
| esv2763570 | Chr.1:81470555 - 81576665 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map