Assay Details
Target Gene Details
Entrez Gene ID: | 8443 |
Gene Name: | glyceronephosphate O-acyltransferase |
Gene Aliases: |
DAP-AT, DAPAT, DHAPAT, RCDP2 |
Location: |
Chr.1:231241173-231277973 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNPAT | NM_001316350.1 | NP_001303279.1 | ||
| NM_014236.3 | NP_055051.1 | |||
| XM_005273313.4 | XP_005273370.1 | |||
| XM_011544303.2 | XP_011542605.1 | |||
| XM_011544304.2 | XP_011542606.1 | |||
| AB209882.1 | BAD93119.1 | |||
| AF043937.1 | AAC24505.1 | |||
| AJ002190.1 | CAA05242.1 | |||
| AK222773.1 | BAD96493.1 | |||
| AK223304.1 | BAD97024.1 | |||
| AK297982.1 | ||||
| AK309994.1 | ||||
| AK311386.1 | ||||
| AK314181.1 | ||||
| BC000450.2 | AAH00450.1 | |||
| DA552987.1 | ||||
| DC366380.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3589106 | Chr.1:231110289 - 231278336 on Build GRCh38 | Gain |
|
| esv3589093 | Chr.1:230482037 - 231277190 on Build GRCh38 | Gain |
|
| nsv1000488 | Chr.1:230799658 - 231269735 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map