Assay Details
Target Gene Details
Entrez Gene ID: | 55970 |
Gene Name: | G protein subunit gamma 12 |
Gene Aliases: |
- |
Location: |
Chr.1:67701466-67833779 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNG12 | NM_018841.5 | NP_061329.3 | ||
| XM_017001809.1 | XP_016857298.1 | |||
| XM_017001810.1 | XP_016857299.1 | |||
| XM_017001811.1 | XP_016857300.1 | |||
| AF119663.1 | AAF17220.1 | |||
| AF123766.1 | AAP97245.1 | |||
| AF365871.1 | AAK53385.1 | |||
| AF493879.1 | AAM12593.1 | |||
| AK223000.1 | BAD96720.1 | |||
| AK293101.1 | ||||
| AL832431.1 | CAH10645.1 | |||
| BC005940.1 | AAH05940.1 | |||
| BP236225.1 | ||||
| BP236699.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs146722403] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map