Assay Details
Target Gene Details
Entrez Gene ID: | 2329 |
Gene Name: | flavin containing monooxygenase 4 |
Gene Aliases: |
FMO2 |
Location: |
Chr.1:171314131-171342084 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FMO4 | NM_002022.2 | NP_002013.1 | ||
| XM_005245045.3 | XP_005245102.1 | |||
| XM_005245046.3 | XP_005245103.1 | |||
| XM_005245048.3 | XP_005245105.1 | |||
| XM_006711243.3 | XP_006711306.1 | |||
| XM_011509347.2 | XP_011507649.1 | |||
| XM_011509348.2 | XP_011507650.1 | |||
| XM_011509349.2 | XP_011507651.1 | |||
| BC002780.2 | AAH02780.1 | |||
| BT007444.1 | AAP36112.1 | |||
| Z11737.1 | CAA77797.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1002423 | Chr.1:171325245 - 171407797 on Build GRCh38 | Loss |
|
| nsv831882 | Chr.1:171197085 - 171356944 on Build GRCh38 | Loss |
|
| esv28731 | Chr.1:171046945 - 171757821 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map