Assay Details
Target Gene Details
Entrez Gene ID: | 155184 |
Gene Name: | solute carrier family 2 member 7 |
Gene Aliases: |
GLUT7 |
Location: |
Chr.1:8992534-9026384 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC2A7 | NM_207420.2 | NP_997303.2 | ||
| XM_011540824.2 | XP_011539126.1 | |||
| AY571960.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545345 | Chr.1:8831066 - 9035551 on Build GRCh38 | Gain |
|
| nsv1004025 | Chr.1:9000983 - 9069086 on Build GRCh38 | Gain |
|
| nsv1009004 | Chr.1:8979645 - 9014406 on Build GRCh38 | Loss |
|
| nsv525092 | Chr.1:8998487 - 9020907 on Build GRCh38 | Gain |
|
| dgv15n100 | Chr.1:8992095 - 9056684 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map