Assay Details
Target Gene Details
Entrez Gene ID: | 378807 |
Gene Name: | cation channel sperm associated 4 |
Gene Aliases: |
- |
Location: |
Chr.1:26189962-26202542 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CATSPER4 | NM_198137.1 | NP_937770.1 | ||
| XM_011541432.2 | XP_011539734.1 | |||
| XM_011541433.2 | XP_011539735.1 | |||
| BC128138.1 | ||||
| BC128139.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1002191 | Chr.1:26149030 - 26219160 on Build GRCh38 | Gain |
|
| esv3577792 | Chr.1:26184821 - 26198215 on Build GRCh38 | Loss |
|
| esv3577794 | Chr.1:26190094 - 26197572 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map