Assay Details
Target Gene Details
Entrez Gene ID: | 6342 |
Gene Name: | sterol carrier protein 2 |
Gene Aliases: |
NLTP, NSL-TP, SCP-2, SCP-CHI, SCP-X, SCPX |
Location: |
Chr.1:52927229-53051617 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCP2 | NM_001007098.2 | NP_001007099.1 | ||
| NM_001193599.1 | NP_001180528.1 | |||
| NM_001193600.1 | NP_001180529.1 | |||
| NM_001193617.1 | NP_001180546.1 | |||
| NM_002979.4 | NP_002970.2 | |||
| XM_005271103.4 | XP_005271160.1 | |||
| XM_011541935.2 | XP_011540237.1 | |||
| XM_017002045.1 | XP_016857534.1 | |||
| AB208789.1 | BAD92026.1 | |||
| AK294631.1 | ||||
| AK295214.1 | ||||
| AK308105.1 | ||||
| AK312856.1 | ||||
| BC067108.1 | AAH67108.1 | |||
| M75883.1 | AAA03557.1 | |||
| M75884.1 | AAA03558.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010121 | Chr.1:52939995 - 53134678 on Build GRCh38 | Gain |
|
| nsv1001535 | Chr.1:52620368 - 53346539 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map