Assay Details
Target Gene Details
Entrez Gene ID: | 5792 |
Gene Name: | protein tyrosine phosphatase, receptor type F |
Gene Aliases: |
BNAH2, LAR |
Location: |
Chr.1:43522238-43623672 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv157n100 | Chr.1:43525952 - 43606749 on Build GRCh38 | Gain |
|
| esv3585869 | Chr.1:43543699 - 43544317 on Build GRCh38 | Loss |
|
| esv2678218 | Chr.1:43543654 - 43544288 on Build GRCh38 | Deletion |
|
| nsv519570 | Chr.1:43539609 - 43606395 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map