Assay Details
Target Gene Details
Entrez Gene ID: | 84871 |
Gene Name: | ATP/GTP binding protein like 4 |
Gene Aliases: |
CCP6 |
Location: |
Chr.1:48520157-50024011 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGBL4 | NM_001323573.1 | NP_001310502.1 | ||
| NM_001323574.1 | NP_001310503.1 | |||
| NM_001323575.1 | NP_001310504.1 | |||
| NM_032785.3 | NP_116174.3 | |||
| NR_136623.1 | ||||
| XM_011542308.2 | XP_011540610.1 | |||
| XM_011542310.2 | XP_011540612.1 | |||
| XM_017002595.1 | XP_016858084.1 | |||
| XM_017002596.1 | XP_016858085.1 | |||
| XM_017002597.1 | XP_016858086.1 | |||
| XM_017002598.1 | XP_016858087.1 | |||
| AK094826.1 | ||||
| AK294394.1 | ||||
| AK294633.1 | ||||
| AK296221.1 |
Target Gene Details
Entrez Gene ID: | 100874313 |
Gene Name: | AGBL4 intronic transcript 1 |
Gene Aliases: |
- |
Location: |
Chr.1:49374201-49472085 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGBL4-IT1 | NR_046839.1 | |||
| AW071649.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003151 | Chr.1:49382052 - 49496947 on Build GRCh38 | Loss |
|
| nsv546194 | Chr.1:49097909 - 49583818 on Build GRCh38 | Loss |
|
| nsv1011949 | Chr.1:49299766 - 49431310 on Build GRCh38 | Loss |
|
| esv2760076 | Chr.1:49387486 - 49533594 on Build GRCh38 | Loss |
|
| esv3577845 | Chr.1:49388770 - 49489029 on Build GRCh38 | Loss |
|
| nsv1006879 | Chr.1:49387474 - 49464868 on Build GRCh38 | Loss |
|
| nsv546196 | Chr.1:49273041 - 49450176 on Build GRCh38 | Loss |
|
| nsv1014234 | Chr.1:49372335 - 49418415 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map