Assay Details
Target Gene Details
Entrez Gene ID: | 23013 |
Gene Name: | spen family transcriptional repressor |
Gene Aliases: |
HIAA0929, MINT, RBM15C, SHARP |
Location: |
Chr.1:15847864-15940455 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPEN | NM_015001.2 | NP_055816.2 | ||
| AF356524.1 | AAK52750.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011969 | Chr.1:15826990 - 15875045 on Build GRCh38 | Gain+Loss |
|
| nsv833647 | Chr.1:15794042 - 15952334 on Build GRCh38 | Loss |
|
| dgv37n100 | Chr.1:15823408 - 15924038 on Build GRCh38 | Gain |
|
| esv2764241 | Chr.1:15827002 - 15855150 on Build GRCh38 | Loss |
|
| nsv3409 | Chr.1:15809988 - 15852694 on Build GRCh38 | Deletion |
|
| dgv38n100 | Chr.1:15839166 - 15924038 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map