Assay Details
Target Gene Details
Entrez Gene ID: | 204962 |
Gene Name: | solute carrier family 44 member 5 |
Gene Aliases: |
CTL5 |
Location: |
Chr.1:75202131-75724261 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC44A5 | NM_001130058.1 | 24 | 2701 | NP_001123530.1 |
| NM_001320285.1 | 26 | 2861 | NP_001307214.1 | |
| NM_001320287.1 | 25 | 2794 | NP_001307216.1 | |
| XM_006710445.3 | 22 | 5474 | XP_006710508.1 | |
| XM_011540986.2 | 23 | 5565 | XP_011539288.1 | |
| XM_011540987.2 | 25 | 5642 | XP_011539289.1 | |
| XM_017000607.1 | 23 | 5613 | XP_016856096.1 | |
| XM_017000608.1 | 23 | 2726 | XP_016856097.1 | |
| XM_017000609.1 | 26 | 6931 | XP_016856098.1 | |
| XM_017000610.1 | 26 | 6009 | XP_016856099.1 | |
| XM_017000611.1 | 24 | 5694 | XP_016856100.1 | |
| AK128063.1 | 25 | 2754 |
More Information
Set Membership: |
|

Genomic Map