Assay Details
Target Gene Details
Entrez Gene ID: | 6646 |
Gene Name: | sterol O-acyltransferase 1 |
Gene Aliases: |
ACACT, ACAT, ACAT-1, ACAT1, SOAT, STAT |
Location: |
Chr.1:179293714-179358680 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOAT1 | NM_001252511.1 | NP_001239440.1 | ||
| NM_001252512.1 | NP_001239441.1 | |||
| NM_003101.5 | NP_003092.4 | |||
| NR_045530.1 | ||||
| XM_011509911.1 | XP_011508213.1 | |||
| XM_011509912.1 | XP_011508214.1 | |||
| AK026611.1 | ||||
| AK290659.1 | ||||
| AK294047.1 | ||||
| AK300551.1 | ||||
| AK310920.1 | ||||
| AK316401.1 | ||||
| AL833625.1 | ||||
| BC028940.1 | AAH28940.1 | |||
| DC344271.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523708 | Chr.1:179322865 - 179324376 on Build GRCh38 | Loss |
|
| nsv1005278 | Chr.1:179228510 - 179402260 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map