Assay Details
Target Gene Details
Entrez Gene ID: | 1806 |
Gene Name: | dihydropyrimidine dehydrogenase |
Gene Aliases: |
DHP, DHPDHASE, DPD |
Location: |
Chr.1:97077743-97921059 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPYD | NM_000110.3 | NP_000101.2 | ||
| NM_001160301.1 | NP_001153773.1 | |||
| XM_005270562.3 | XP_005270619.2 | |||
| XM_006710397.3 | XP_006710460.1 | |||
| XM_017000507.1 | XP_016855996.1 | |||
| XM_017000508.1 | XP_016855997.1 | |||
| XM_017000509.1 | XP_016855998.1 | |||
| XM_017000510.1 | XP_016855999.1 | |||
| AB003063.1 | BAA89789.1 | |||
| AK291217.1 | ||||
| AK297515.1 | ||||
| BC008379.1 | AAH08379.1 | |||
| BC064027.1 | AAH64027.1 | |||
| BC108742.1 | AAI08743.1 | |||
| BC131777.1 | ||||
| BC131778.1 | ||||
| BT006740.1 | AAP35386.1 | |||
| DA721123.1 | ||||
| U09178.1 | AAA57474.1 | |||
| U20938.1 | AAB51366.1 |
Target Gene Details
Entrez Gene ID: | 100873933 |
Gene Name: | DPYD antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.1:97796921-97798051 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPYD-AS2 | NR_046591.1 | 1 | 87 | |
| BU569038.1 | 1 | 46 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv376n54 | Chr.1:97787849 - 97871727 on Build GRCh38 | Loss |
|
| nsv830725 | Chr.1:97792338 - 97977941 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map