Assay Details
Target Gene Details
Entrez Gene ID: | 91624 |
Gene Name: | nexilin F-actin binding protein |
Gene Aliases: |
CMH20, NELIN |
Location: |
Chr.1:77888515-77946329 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEXN | NM_001172309.1 | NP_001165780.1 | ||
| NM_144573.3 | NP_653174.3 | |||
| XM_005271322.3 | XP_005271379.1 | |||
| XM_005271323.3 | XP_005271380.1 | |||
| XM_005271324.4 | XP_005271381.1 | |||
| XM_005271325.3 | XP_005271382.1 | |||
| XM_005271326.3 | XP_005271383.1 | |||
| XM_005271327.3 | XP_005271384.1 | |||
| AF114264.1 | AAD29607.1 | |||
| AF176780.1 | AAQ13658.1 | |||
| AK057954.1 | BAB71622.1 | |||
| AK293201.1 | ||||
| AK298565.1 | ||||
| AK300728.1 | ||||
| AK307487.1 | ||||
| BC017827.1 | ||||
| BC055084.1 | AAH55084.1 | |||
| BC111395.1 | ||||
| BC114444.1 | ||||
| BC114445.1 | ||||
| BX648546.1 | ||||
| DQ464902.1 | ||||
| S67069.1 | AAB28815.1 |
More Information
Set Membership: |
|

Genomic Map