Assay Details
Target Gene Details
Entrez Gene ID: | 388630 |
Gene Name: | TraB domain containing 2B |
Gene Aliases: |
TIKI2 |
Location: |
Chr.1:47760528-47997367 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRABD2B | NM_001194986.1 | 7 | 2521 | NP_001181915.1 |
| XM_006710640.3 | 8 | 3102 | XP_006710703.1 | |
| XM_017001260.1 | 7 | 2832 | XP_016856749.1 | |
| XM_017001261.1 | 5 | 2552 | XP_016856750.1 | |
| XM_017001262.1 | XP_016856751.1 | |||
| XM_017001263.1 | 5 | 2971 | XP_016856752.1 | |
| AK090467.1 | BAC03448.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829760 | Chr.1:47645454 - 47842516 on Build GRCh38 | Loss |
|
| nsv1001126 | Chr.1:47698950 - 47855190 on Build GRCh38 | Gain |
|
| nsv1003161 | Chr.1:47742043 - 47820891 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map