Assay Details
Target Gene Details
Entrez Gene ID: | 2773 |
Gene Name: | G protein subunit alpha i3 |
Gene Aliases: |
87U6, ARCND1 |
Location: |
Chr.1:109548564-109595843 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNAI3 | NM_006496.3 | NP_006487.1 | ||
| AF493907.1 | AAM12621.1 | |||
| AK311691.1 | ||||
| AK312252.1 | ||||
| BC025285.1 | AAH25285.1 | |||
| BT019973.1 | AAV38776.1 | |||
| BT019974.1 | AAV38777.1 | |||
| J03005.1 | AAA52557.1 | |||
| J03198.1 | AAA35896.1 | |||
| J03238.1 | AAA35939.1 | |||
| M27543.1 | AAA52579.1 |
More Information
Set Membership: |
|

Genomic Map