Assay Details
Target Gene Details
Entrez Gene ID: | 6533 |
Gene Name: | solute carrier family 6 member 6 |
Gene Aliases: |
TAUT |
Location: |
Chr.3:14402576-14489349 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC6A6 | NM_001134367.3 | NP_001127839.2 | ||
| NM_001134368.3 | NP_001127840.1 | |||
| NM_003043.5 | NP_003034.2 | |||
| NR_103507.2 | ||||
| XM_011534031.2 | XP_011532333.1 | |||
| AB209172.1 | BAD92409.1 | |||
| AK303649.1 | ||||
| AK307284.1 | ||||
| AK308417.1 | ||||
| AK309534.1 | ||||
| BC006252.2 | AAH06252.1 | |||
| BC111489.1 | AAI11490.1 | |||
| BC137128.1 | ||||
| BC137129.1 | ||||
| BM149397.1 | ||||
| DC359099.1 | ||||
| U16120.1 | AAA50842.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834619 | Chr.3:14354326 - 14487969 on Build GRCh38 | Gain+Loss |
|
| esv2724945 | Chr.3:13931758 - 14820330 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map