Assay Details
Target Gene Details
Entrez Gene ID: | 152110 |
Gene Name: | NIMA related kinase 10 |
Gene Aliases: |
- |
Location: |
Chr.3:27107336-27369460 on Build GRCh38 |
Assay Gene Location: | Within Exon 47 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEK10 | XM_006712997.3 | 38 | 4873 | XP_006713060.1 |
| XM_006712999.3 | XP_006713062.1 | |||
| XM_006713001.3 | 37 | 4732 | XP_006713064.1 | |
| XM_017005761.1 | 38 | 4855 | XP_016861250.1 | |
| XM_017005762.1 | 40 | 5019 | XP_016861251.1 | |
| XM_017005763.1 | 39 | 5000 | XP_016861252.1 | |
| XM_017005764.1 | 38 | 4832 | XP_016861253.1 | |
| XM_017005765.1 | 38 | 4804 | XP_016861254.1 | |
| XM_017005766.1 | 37 | 4825 | XP_016861255.1 | |
| XM_017005767.1 | 37 | 4768 | XP_016861256.1 | |
| XM_017005768.1 | 38 | 4765 | XP_016861257.1 | |
| XM_017005769.1 | 38 | 4762 | XP_016861258.1 | |
| XM_017005770.1 | 37 | 4714 | XP_016861259.1 | |
| XM_017005771.1 | 36 | 4684 | XP_016861260.1 | |
| XM_017005772.1 | 36 | 4627 | XP_016861261.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv880e214 | Chr.3:26983690 - 27243989 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map