Assay Details
Target Gene Details
Entrez Gene ID: | 55164 |
Gene Name: | SHQ1, H/ACA ribonucleoprotein assembly factor |
Gene Aliases: |
GRIM-1, Shq1p |
Location: |
Chr.3:72725703-72848475 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHQ1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8375n54 | Chr.3:72724215 - 72735474 on Build GRCh38 | Loss |
|
| nsv590631 | Chr.3:72732656 - 72738435 on Build GRCh38 | Loss |
|
| dgv8377n54 | Chr.3:72730895 - 72735474 on Build GRCh38 | Loss |
|
| esv2763278 | Chr.3:72595871 - 73026595 on Build GRCh38 | Gain |
|
| nsv954887 | Chr.3:72731150 - 72739849 on Build GRCh38 | Deletion |
|
| esv28177 | Chr.3:72730836 - 72736909 on Build GRCh38 | Loss |
|
| esv3596544 | Chr.3:72730924 - 72736453 on Build GRCh38 | Gain |
|

Genomic Map