Assay Details
Target Gene Details
Entrez Gene ID: | 131405 |
Gene Name: | tripartite motif containing 71 |
Gene Aliases: |
LIN-41, LIN41 |
Location: |
Chr.3:32818018-32897826 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIM71 | NM_001039111.2 | NP_001034200.1 | ||
| DQ232881.1 | ABB18374.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003285 | Chr.3:31386933 - 33172886 on Build GRCh38 | Gain |
|
| nsv998302 | Chr.3:32595752 - 33112754 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76312503] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map