Assay Details
Target Gene Details
Entrez Gene ID: | 101928583 |
Gene Name: | uncharacterized LOC101928583 |
Gene Aliases: |
- |
Location: |
Chr.3:170467285-170755205 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928583 | NR_135555.1 | |||
| NR_135556.1 | ||||
| NR_135557.1 | ||||
| HY157714.1 |
Target Gene Details
Entrez Gene ID: | 57709 |
Gene Name: | solute carrier family 7 member 14 |
Gene Aliases: |
PPP1R142 |
Location: |
Chr.3:170459554-170586074 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC7A14 | NM_020949.2 | NP_066000.2 | ||
| AB046833.1 | BAB13439.1 | |||
| AK122655.1 | ||||
| BC022968.1 |
More Information
Set Membership: |
|

Genomic Map