Assay Details
Target Gene Details
Entrez Gene ID: | 56852 |
Gene Name: | RAD18, E3 ubiquitin protein ligase |
Gene Aliases: |
RNF73 |
Location: |
Chr.3:8877196-8969249 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD18 | NM_020165.3 | NP_064550.3 | ||
| XM_017006873.1 | XP_016862362.1 | |||
| AB035274.2 | BAA99284.1 | |||
| AF169796.1 | AAF80856.1 | |||
| AK023075.1 | BAB14392.1 | |||
| AK222771.1 | BAD96491.1 | |||
| AY004333.1 | AAF86618.1 | |||
| BC001302.1 | AAH01302.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv275011 | Chr.3:8908509 - 8918634 on Build GRCh38 | Gain+Loss |
|
| nsv518352 | Chr.3:8905994 - 9089915 on Build GRCh38 | Loss |
|
| nsv520776 | Chr.3:8913275 - 8947125 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map