Assay Details
Target Gene Details
Entrez Gene ID: | 64084 |
Gene Name: | calsyntenin 2 |
Gene Aliases: |
ALC-GAMMA, CDHR13, CS2, CSTN2, alcagamma |
Location: |
Chr.3:139935185-140577397 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLSTN2 | NM_022131.2 | NP_071414.2 | ||
| AJ278018.1 | CAC14924.1 | |||
| AK096432.1 | ||||
| AK315308.1 | ||||
| AY753303.1 | AAV30553.1 | |||
| BC103496.1 | AAI03497.1 | |||
| BC103506.1 | AAI03507.1 | |||
| BC104485.1 | AAI04486.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv998423 | Chr.3:139807604 - 140228494 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map