Assay Details
Target Gene Details
Entrez Gene ID: | 28976 |
Gene Name: | acyl-CoA dehydrogenase family member 9 |
Gene Aliases: |
NPD002 |
Location: |
Chr.3:128879490-128916075 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACAD9 | NM_014049.4 | NP_054768.2 | ||
| NR_033426.1 | ||||
| XM_017006264.1 | XP_016861753.1 | |||
| AB209427.1 | 2 | 1312 | BAD92664.1 | |
| AF327351.1 | AAL56011.1 | |||
| AK022568.1 | BAB14104.1 | |||
| AK024012.1 | BAB14775.1 | |||
| AK130311.1 | ||||
| AK294597.1 | ||||
| AK297952.1 | ||||
| AK310644.1 | ||||
| BC007970.2 | AAH07970.1 | |||
| BC013354.2 | AAH13354.1 | |||
| DA630022.1 |
More Information
Set Membership: |
|

Genomic Map