Assay Details
Target Gene Details
Entrez Gene ID: | 100126791 |
Gene Name: | eosinophil granule ontogeny transcript (non-protein coding) |
Gene Aliases: |
EGO, NCRNA00190 |
Location: |
Chr.3:4749192-4751590 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EGOT | NR_004428.1 | 1 | 58 |
Target Gene Details
Entrez Gene ID: | 3708 |
Gene Name: | inositol 1,4,5-trisphosphate receptor type 1 |
Gene Aliases: |
ACV, CLA4, INSP3R1, IP3R, IP3R1, PPP1R94, SCA15, SCA16, SCA29 |
Location: |
Chr.3:4493348-4847840 on Build GRCh38 |
Assay Gene Location: | Within Intron 45 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv460357 | Chr.3:4656425 - 4751836 on Build GRCh38 | Loss |
|
| nsv1005718 | Chr.3:4722315 - 5046200 on Build GRCh38 | Gain |
|
| esv3893643 | Chr.3:4529987 - 4974421 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map