Assay Details
Target Gene Details
Entrez Gene ID: | 27086 |
Gene Name: | forkhead box P1 |
Gene Aliases: |
12CC4, HSPC215, MFH, QRF1, hFKH1B |
Location: |
Chr.3:70954714-71583989 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Target Gene Details
Entrez Gene ID: | 104502416 |
Gene Name: | FOXP1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:71289758-71305854 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXP1-AS1 | NR_126463.1 | |||
| AA399395.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv999942 | Chr.3:71131662 - 71736899 on Build GRCh38 | Gain |
|
| nsv1010076 | Chr.3:71290647 - 71337382 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map