Assay Details
Target Gene Details
Entrez Gene ID: | 8419 |
Gene Name: | beaded filament structural protein 2 |
Gene Aliases: |
CP47, CP49, CTRCT12, LIFL-L, PHAKOSIN |
Location: |
Chr.3:133397882-133475212 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BFSP2 | NM_003571.3 | NP_003562.1 | ||
| XM_017007315.1 | XP_016862804.1 | |||
| BC113518.1 | ||||
| BC113520.1 | ||||
| BM696550.1 | ||||
| U48224.1 | AAC50414.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8607n54 | Chr.3:133413620 - 133418440 on Build GRCh38 | Gain |
|
| nsv591819 | Chr.3:133415190 - 133418440 on Build GRCh38 | Gain |
|
| nsv955348 | Chr.3:133414057 - 133418456 on Build GRCh38 | Deletion |
|
| nsv591820 | Chr.3:133415517 - 133424474 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map