Assay Details
Target Gene Details
Entrez Gene ID: | 26137 |
Gene Name: | zinc finger and BTB domain containing 20 |
Gene Aliases: |
DPZF, HOF, ODA-8S, PRIMS, ZNF288 |
Location: |
Chr.3:114314500-115147280 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB20 | NM_001164342.2 | 5 | 25732 | NP_001157814.1 |
| NM_001164343.2 | 12 | 26194 | NP_001157815.1 | |
| NM_001164344.2 | 8 | 26158 | NP_001157816.1 | |
| NM_001164345.2 | 7 | 26112 | NP_001157817.1 | |
| NM_001164346.2 | 6 | 25751 | NP_001157818.1 | |
| NM_001164347.2 | 6 | 25773 | NP_001157819.1 | |
| NM_015642.5 | 10 | 25956 | NP_056457.3 | |
| NR_121662.1 | 7 | 24214 | ||
| AL049423.1 | 1 | 502 |
More Information
Set Membership: |
|

Genomic Map